MilliporeSigma
  • Possible involvement of lipoxygenase products of arachidonic acid pathway in ovulation.
Possible involvement of lipoxygenase products of arachidonic acid pathway in ovulation.
Prostaglandins (1983-12-01)
R Reich, F Kohen, Z Naor, A Tsafriri
ABSTRACT
The possible involvement of products of the lipoxygenase pathway of arachidonic acid cascade in ovulation was tested by intrabursal injection of nordihydroguaiaretic acid (NDGA); 5, 8, 11-eicosatriynoic acid (5, 8, 11-ETYA), 3 amino-1-(3 trifluromethyphenyl)-2-pyrazoline hydrochloride (BW755c) and (FPL 55712). All these drugs reduced the number of ova released from the treated ovaries in a dose-dependent manner, without affecting ovulation from contralateral ovaries. NDGA was most potent since it completely blocked ovulation from the treated ovaries in 17/38 rats receiving a dose higher than 0.15 mg/bursa. This effect of NDGA cannot be ascribed to its inhibition of ovarian PGE synthesis. Conversion of labeled arachidonic acid via the lipoxygenase pathway by preovulatory rat follicles was demonstrated by TLC chromatography. Collectively, these results suggest the involvement of products of lipoxygenase pathway of arachidonic acid in ovulation in the rat.