MilliporeSigma
  • [Effect of 2-amino-2-thiazoline on the sulfhydryl group content of yeasts of various degrees of radiosensitivity].
[Effect of 2-amino-2-thiazoline on the sulfhydryl group content of yeasts of various degrees of radiosensitivity].
Nauchnye doklady vysshei shkoly. Biologicheskie nauki (1984-01-01)
L T Zolotareva, E E Graevskaia, E N Goncharenko
ABSTRACT
The influence of 2-amino-2-thiazoline on the level of total sulphydryl groups in the haploid and diploid yeast cells Saccharomyces cerevisiae and Pichia guilliermondii has been studied. The increase of radioresistance of the yeast cells has been shown to be connected with the growing content of the endogenous thiols.