MilliporeSigma
  • Cytochemical characterization of pituitary target cells for biotinylated gonadotropin releasing hormone.
Cytochemical characterization of pituitary target cells for biotinylated gonadotropin releasing hormone.
Peptides (1983-07-01)
G V Childs, Z Naor, E Hazum, R Tibolt, K N Westlund, M B Hancock
ABSTRACT
These studies describe the application of new cytochemical stains that co-localize a biotin-labeled gonadotropin releasing hormone (GnRH) analog and FSH or LH in the same field or cell. Pituitary monolayer cells were stimulated with the [D-Lys6] GnRH analog or the same analog labeled with biotin. Biotinylated [D-Lys6] GnRH exhibited a higher affinity and was 7-10 X more potent than unlabeled [D-Lys6] GnRH. The avidin-biotin peroxidase complex technique (ABC) was applied to localize the biotinylated GnRH on the cells with the use of a dense black peroxidase substrate. Specificity tests showed that the stain could be eliminated by competition with unlabeled [D-Lys6] GnRH. The GnRH stain was followed by immunocytochemical stains for LH beta, FSH beta or 25-39ACTH with a different peroxidase substrate (amber or orange-red). Stain for GnRH was found on the surfaces of 16% of the cells and 60-90% of the GnRH stained cells also stained for one of the gonadotropins. Most (90-100%) of the gonadotropes showed stain for GnRH. Our studies demonstrate that a potent biotinylated GnRH analog binds cells that can be identified specifically as gonadotropes.