MilliporeSigma
  • Corneal penetration and metabolism of the antiherpetic 5-iodo-2'-deoxycytidine.
Corneal penetration and metabolism of the antiherpetic 5-iodo-2'-deoxycytidine.
Current eye research (1985-05-01)
D Sincholle, J P Conduzorgues, B Mbatchi, J P Masse
ABSTRACT
The study of the permeability of the cornea to 5-iodo-2'-deoxycytidine (IDC), an antiherpetic agent was performed in the rabbit. In a first experiment, using 125I-IDC eye-drops and a sustained contact between the drug and the cornea, we showed that the penetration of IDC in the aqueous humor was important. In a second experiment, using a HPLC method, we studied comparative ocular penetration and metabolism of IDC and 5-iodo-2'-deoxyuridine (IDU).