MilliporeSigma
  • Highly sensitive solid-phase immunoenzymometric assay for placental and placental-like alkaline phosphatases with a monoclonal antibody and monodisperse polymer particles.
Highly sensitive solid-phase immunoenzymometric assay for placental and placental-like alkaline phosphatases with a monoclonal antibody and monodisperse polymer particles.
Clinical chemistry (1985-01-01)
J L Millán, K Nustad, B Nørgaard-Pedersen
ABSTRACT
We have used a mouse monoclonal antibody (H7) to placental alkaline phosphatase (PLAP, EC 3.1.3.1) in developing an immunoenzymometric assay for PLAP and PLAP-like enzymes. The antibody is bound to sheep anti-mouse IgG (Ab2) covalently coupled to tosylated shell-and-core light (1.07 g/cm3) monodisperse polymer particles. Adding the H7-Ab2-polymer particle suspension to a PLAP-containing sample gives maximal binding of the antigen within 10 min. PLAP and PLAP-like enzymes remain active and bound to the solid-phase throughout all assay manipulations, and can thus be saved for future testing. In testing the enzymes for inhibition by L-Phe, L-Phe-Gly-Gly, L-Leu, and L-homoarginine, the effect of all the inhibitors is fully reversible. The assay is highly versatile, and its sensitivity (routinely 0.05 micrograms/L) can be increased 1000-fold by adjusting the sample volume and incubation time (sample volume is irrelevant between 50 microL and 5 mL). We have measured the basal activities of PLAP in men and women and, by using enzyme inhibitors, have characterized it as corresponding to the PLAP-like phenotypes described in normal human testis.
Sigma-Aldrich
Phe-Gly-Gly