MilliporeSigma
  • [Prevention of post-resuscitation impairment of energy metabolism in the myocardium using sodium gamma-hydroxybutyrate, ionol and gutimine].
[Prevention of post-resuscitation impairment of energy metabolism in the myocardium using sodium gamma-hydroxybutyrate, ionol and gutimine].
Voprosy meditsinskoi khimii (1988-01-01)
V T Dolgikh
ABSTRACT
Long-term impairments of energy metabolisms in myocardium were found in rats after clinical death caused by acute hemorrhage. Bioenergetics of myocardium tended to normalize within a month after resuscitation. Severity of postresuscitation period correlated with the level of deteriorations observed in energy metabolism. Preadministration of gamma-aminobutyric acid, ionol and gutimine decreased these impairments as well as the postresuscitation lethality. Considering the final results of resuscitation the efficiency of the drugs studied might be arranged in a following order: gamma-aminobutyric acid, ionol, gutimine.
Sigma-Aldrich
2-Imino-4-thiobiuret, 99%