MilliporeSigma
  • Converting-enzyme inhibitor administration lowers urinary free 19-nor-deoxycorticosterone levels.
Converting-enzyme inhibitor administration lowers urinary free 19-nor-deoxycorticosterone levels.
Hypertension (Dallas, Tex. : 1979) (1985-03-01)
G T Griffing, T E Wilson, J C Melby
ABSTRACT
19-Nor-deoxycorticosterone (19-nor-DOC) is a human mineralocorticoid. The regulation of its secretion is poorly understood, as renin angiotensin II (ANG II) stimulation has minimal effects on 19-nor-DOC. This study sought to determine if ANG II inhibition would decrease 19-nor-DOC production. Six normal subjects on fixed electrolyte intake were admitted to a metabolic unit. After a 5-day control period to establish electrolyte balance, enalapril, p.o., 10 mg/day, was administered for 28 days. This treatment resulted in ANG II inhibition, which was reflected by a rise in plasma renin activity, a blunting of the postural plasma aldosterone increment, and a decrease in aldosterone secretion rate (ASR). Levels of urinary free (UF) 19-nor-DOC progressively decreased from 294 +/- 108 ng/day on Day 0 to 164 +/- 70 on Day 3, 141 +/- 62 on Day 7, 101 +/- 38 on Day 14, 68 +/- 18 on Day 21, and 106 +/- 31 on Day 28. The decrease in 19-nor-DOC levels was synchronous with the fall in ASR (R = 0.94, n = 5, p less than 0.005), but it was of greater magnitude (71% decrease in 19-nor-DOC levels versus 41% decrease in ASR). In addition, the decrease in 19-nor-DOC levels correlated with a fall in urinary potassium and an increase in both urinary sodium and chloride (R = 0.68, -0.79, -0.87 respectively; n = 6, p less than 0.05). The fall in ASR, on the other hand, was not significantly correlated with the changes in these urinary electrolyte levels (R = 0.65, 0.64, 0.57 respectively; n = 5).(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
3α,5β-Tetrahydroaldosterone, ≥98% (CP)
Pricing and availability is not currently available.