MilliporeSigma
Facilitation of insulin release by N-p-tosylglycine.
Biochemical pharmacology (1985-07-15)
A Sener, R Gomis, B Billaudel, W J Malaisse
ABSTRACT
N-p-tosylglycine, which inhibits transglutaminase activity in islet homogenates, was found to cause a rapid and sustained facilitation of insulin release evoked by D-glucose, L-leucine or the association of Ba2+ and theophylline in intact islets. Such a facilitating action could not be attributed to any obvious effect upon either nutrient oxidation or 45Ca net uptake and outflow. It failed to be reproduced by glycine, N alpha-p-tosyl-L-arginine methyl ester or N alpha-p-tosyl-L-lysine methyl ester. N-p-tosylglycine (5.0 mM) slightly enhanced insulin release evoked by a high concentration of glucose (16.7 mM) and failed to affect significantly the secretory response to the association of L-leucine and L-glutamine or that of D-glucose and gliclazide. N-p-tosylglycine failed to affect the incorporation of [2,5-3H]histamine in trichloroacetic acid-precipitable material in intact islets. These results suggest that N-p-tosylglycine interferes with a late event in the secretory sequence, possibly at the level of the cell boundary, rather than inhibiting the crosslinking of intracellular proteins.