MilliporeSigma
  • High performance liquid chromatographic quantitation of carboxypeptidase activity secreted by human hep G2 cells.
High performance liquid chromatographic quantitation of carboxypeptidase activity secreted by human hep G2 cells.
Analytical biochemistry (1988-04-01)
B G Grimwood, A L Tarentino, T H Plummer
ABSTRACT
A new high performance liquid chromatographic method has been developed for the determination of carboxypeptidase N activity which quantitates the furylacryloyl-alanine released by enzymatic cleavage of furylacryloyl-alanyl-lysine or furylacryloyl-alanyl-arginine. A short isocratic gradient elutes the substrate and product in less than 7 min and multiple analyses are facilitated by an automatic sample injector. The microassay readily detects and quantitates carboxypeptidase N activity secreted into culture medium. It was determined that approximately 1 X 10(6) Hep G2 cells at early confluence secreted 1 ng of carboxypeptidase N in 24 h. The microassay will also detect as little as 51 pg of purified carboxypeptidase N or 8 pg of carboxypeptidase B.
Sigma-Aldrich
N-(3-[2-Furyl]acryloyl)-Ala-Lys, powder
Pricing and availability is not currently available.