MilliporeSigma
[Conformational analysis of 5-substituted uracil].
Biofizika (1989-09-01)
S G Stepan'ian, E D Radchenko, G G Sheina, Iu P Vlagoĭ
ABSTRACT
The high-resolution IR-spectra of 5-nitrouracil and 5-bromouracil isolated in Ar matrices at 11 K were obtained for the first time. The conformational structure of uracil 5-substituents--thymine, 5-bromouracil, 5-nitrouracil--is calculated by the molecular mechanics and quantum--chemical MINDO/3 methods. The possibility of thymine transition to nonplanar conformations is observed.