MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Analysis of inhibitors of the site-specific recombination reaction mediated by Tn3 resolvase.
Analysis of inhibitors of the site-specific recombination reaction mediated by Tn3 resolvase.
Journal of molecular biology (1989-03-20)
P M Flanagan, M A Fennewald
ABSTRACT
The Tn3-encoded resolvase protein promotes a site-specific recombination reaction between two directly repeated copies of the recombination site res. Several inhibitors that block this event in vitro have been isolated. In this study four of these inhibitors were tested on various steps in the recombination reaction. Two inhibitors. A9387 and A1062, inhibit resolvase binding to the res site. Further, DNase I footprinting revealed that at certain concentrations of A9387 and A1062, resolvase was preferentially bound to site I of res, the site containing the recombinational crossover point. The two other inhibitors, A20812 and A21960, do not affect resolvase binding and bending of the DNA but inhibit synapse formation between resolvase and two directly repeated res sites.