MilliporeSigma
  • Phase II clinical trial of high-dose toremifene as primary hormone therapy in aromatase inhibitor-resistant breast cancer.
Phase II clinical trial of high-dose toremifene as primary hormone therapy in aromatase inhibitor-resistant breast cancer.
Gan to kagaku ryoho. Cancer & chemotherapy (2013-07-19)
Hideaki Ogata, Yasushi Okamoto, Youichi Arima, Hisaki Fukushima, Hiroshi Takeyama, Akinori Yamashita, Masao Kinoshita, Noriyuki Suzuki, Terumasa Sawada, Yoshitomo Koshida, Akira Matsui, Akihiko Tachibana, Hiroshi Nakayama, Yoko Oishi, Hiroko Nogi, Ken Uchida
ABSTRACT
Third-generation aromatase inhibitors(AIs)are now common in adjuvant hormone therapy for breast cancer in postmenopausal women. However, a suitable treatment has yet to be established for patients who develop cancer recurrence during or after adjuvant AI therapy. This prospective study evaluated the efficacy and safety of 120mg/day toremifene citrate(TOR-120)administered orally to 23 patients with recurrent breast cancer who were receiving or had received adjuvant AI therapy. Primary therapy for recurrence was TOR-120 monotherapy. The response rate was 13. 0%(partial response: three patients), the clinical benefit rate was 78. 3%(partial response: three patients; long-term stable disease: 15 patients), and median time to progression was 8. 1 months. Grade 1 adverse events such as loss of appetite, sweating, flushing and edema face were observed. TOR-120 monotherapy was effective and safe as a primary hormone therapy for recurrent breast cancer unresponsive to AIs.
Sigma-Aldrich
Toremifene citrate salt, ≥98% (HPLC)