MilliporeSigma
Effect of nitric oxide on anterior segment physiology in monkeys.
Investigative ophthalmology & visual science (2013-06-27)
Galen W Heyne, Julie A Kiland, Paul L Kaufman, B'Ann T Gabelt
ABSTRACT
To determine the effect of the nitric oxide donor, sodium nitroprusside (SNP), and the nitric oxide synthase (NOS) inhibitor, L-nitro-arginine-methylester (L-NAME), on IOP, mean arterial pressure (MAP), pupil diameter (PD), refraction (Rfx), aqueous humor formation (AHF), and outflow facility (OF) in monkeys. Monkeys were treated with single or multiple topical treatments of 500 μg SNP or L-NAME to one eye. IOP was determined by Goldmann applanation tonometry, PD with vernier calipers in room light, Rfx by Hartinger coincidence refractometry, AHF by fluorophotometry, and MAP with a blood pressure monitor. OF was determined by two-level constant pressure perfusion following anterior chamber exchange. Following four topical treatments with 500 μg SNP, 30 minutes apart, IOP was significantly decreased from 2 to 6 hours compared with the contralateral control with the maximum IOP reduction of 20% at 3 hours (P < 0.001). PD, Rfx, and AHF were unchanged. Effects on MAP were variable. OF after SNP exchange was significantly increased by 77% (P < 0.05) at 10(-3) M. Topical L-NAME had no effect on IOP, PD, Rfx, or MAP. Enhancement of nitric oxide concentration at targeted tissues in the anterior segment may be a useful approach for IOP reduction for glaucoma therapy. Additional studies are warranted before conclusions can be made regarding the effect of NOS inhibition on ocular physiology in nonhuman primates.
Sigma-Aldrich
Sodium nitroferricyanide(III) dihydrate, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitroferricyanide(III) dihydrate, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitroprusside dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Pricing and availability is not currently available.