MilliporeSigma
  • Analysis of Salvinorin A in urine using microextraction in packed syringe and GC-MS/MS.
Analysis of Salvinorin A in urine using microextraction in packed syringe and GC-MS/MS.
Bioanalysis (2013-03-15)
Ivo Moreno, Beatriz da Fonseca, David Oppolzer, Ana Martinho, Mário Barroso, Angelines Cruz, João António Queiroz, Eugenia Gallardo
ABSTRACT
The aim of this work was to develop and validate a method for the determination of Salvinorin A in human urine using microextraction by packed sorbent (MEPS) and GC-MS/MS. The technique uses a sample volume as low as 0.2 ml, and the analyte was extracted using a C18 sorbent. The method showed to be linear between 20 and 1000 ng/ml and presented a LOD of 5 ng/ml. Intra- and inter-day precision and accuracy were acceptable. Absolute recoveries ranged from 71 to 80%. GC-MS/MS with MEPS demonstrated to be a fast and simple procedure for the quantification of Salvinorin A in urine. This is the first time that GC-MS/MS with MEPS was used for the determination of this compound in biological fluids. Furthermore, the device could be reused for up to 80 extractions, which accounted for a lower cost of analysis.
Sigma-Aldrich
Salvinorin A, ≥98% (HPLC), solid