MilliporeSigma
  • Role of antisecretory agents for gastric endoscopic submucosal dissection.
Role of antisecretory agents for gastric endoscopic submucosal dissection.
Digestive endoscopy : official journal of the Japan Gastroenterological Endoscopy Society (2013-02-02)
Mitsuhiro Fujishiro, Philip W Y Chiu, Hsui-Po Wang
ABSTRACT
Gastric endoscopic submucosal dissection (ESD) causes artificial gastric ulcers and there is no consensus regarding the optimal perioperative management in terms of prevention of intra- or postoperative bleeding and promotion of healing. Traditionally, 8-week administration of proton pump inhibitors (PPI) and mucosal protective agents were used in the same way as for peptic ulcer management. However, recent studies have revealed that prior use of PPI might reduce intraoperative bleeding or early-phase postoperative bleeding, and combination of histamine-2 receptor antagonist (H2RA), and second-look endoscopy might have a similar effect on postoperative bleeding to PPI. Additionally, the advantage of PPI over H2RA is not proven and the optimal duration of PPI may be shortened until 2 weeks when the deteriorating factors for ESD ulcer are excluded. Furthermore, mucosal protective agents may facilitate ulcer healing. Further studies are needed to determine the optimal treatment protocol before and after ESD for both prevention of bleeding complication and promotion of ulcer healing, by using available antisecretory agents and mucosal protective agents.
Sigma-Aldrich
Sucrose octasulfate–aluminum complex