MilliporeSigma
  • Silver(I) and copper(I) adducts of a tris(pyrazolyl)borate decorated with nine trifluoromethyl groups.
Silver(I) and copper(I) adducts of a tris(pyrazolyl)borate decorated with nine trifluoromethyl groups.
Inorganic chemistry (2013-02-02)
Naleen B Jayaratna, Igor I Gerus, Roman V Mironets, Pavel K Mykhailiuk, Muhammed Yousufuddin, H V Rasika Dias
ABSTRACT
Silver and copper ethylene adducts and the silver carbonyl complex of the tris(pyrazolyl)borate [HB(3,4,5-(CF(3))(3)Pz)(3)](-) (which is based on one of the most acidic pyrazoles known) have been synthesized. (13)C NMR resonance signals of metal-bound ethylene carbon atoms of [HB(3,4,5-(CF(3))(3)Pz)(3)]Ag(C(2)H(4)) and [HB(3,4,5-(CF(3))(3)Pz)(3)]Cu(C(2)H(4)) appear at δ 111.6 and 94.9, respectively. The CO stretching frequency of the silver adduct [HB(3,4,5-(CF(3))(3)Pz)(3)]Ag(CO) is significantly higher than that of free CO, but it appears at a region less sensitive to the ligand electronic effects of tris(azolyl)borate silver adducts.