MilliporeSigma
  • Facile synthesis of AgInS₂ hierarchical flowerlike nanoarchitectures composed of ultrathin nanowires.
Facile synthesis of AgInS₂ hierarchical flowerlike nanoarchitectures composed of ultrathin nanowires.
Nanoscale (2013-01-19)
Zhongping Liu, Kaibin Tang, Dake Wang, Linlin Wang, Qiaoyan Hao
ABSTRACT
In this work, AgInS(2) hierarchical flowerlike nanoarchitectures, which are composed of ultrathin nanowires, were synthesized by thermolysis of a mixed solution of AgNO(3), InCl(3)·4H(2)O and n-dodecanethiol at elevated temperature. The average diameter and length of the nanowires composing the nanoarchitectures can reach 5 nm and ∼300 nm, respectively. We investigated the growth process of the nanoarchitectures and the effects of reaction parameters by XRD, SEM and TEM. In particular, the use of InCl(3)·4H(2)O played a decisive role in the synthesis of the nanoarchitectures. Moreover, it was found that polyhedra formed in the initial reaction time, and then the nanowires grew on the facets of these polyhedra, which resulted in the nanoarchitectures. The reaction temperature and the concentration of metal salts could influence the size of the nanowires.
Sigma-Aldrich
Silver sulfide, 99.9% trace metals basis