MilliporeSigma
  • Spectroscopic (FT-IR/FT-Raman) and computational (HF/DFT) investigation and HOMO/LUMO/MEP analysis on 2-amino-4-chlorophenol.
Spectroscopic (FT-IR/FT-Raman) and computational (HF/DFT) investigation and HOMO/LUMO/MEP analysis on 2-amino-4-chlorophenol.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-01)
S Ramalingam, S Periandy, M Karabacak, N Karthikeyan
ABSTRACT
The spectra (FT-IR and FT-Raman) of the present compound; 2-amino-4-chlorophenol (2A4CP) were recorded in the range of 4000-100 cm(-1). All the computational calculations were made in the ground state using the HF and DFT (B3LYP and B3PW91) methods with 6-31++G(d,p) and 6-311++G(d,p) basis sets. From potential energy surface calculation, there are two conformers, Rot-1 and Rot-2 for this molecule. The computational results detected that Rot-1 form is the most stable conformer. Making use of the recorded data, the complete vibrational assignments were made and analysis of the observed fundamental bands of molecule is carried out. The complete assignments were performed on the basis of the total energy distribution (TED) of the vibrational modes, calculated with scaled quantum mechanics (SQMs) method and PQS program. The shifting of the frequencies in the vibrational pattern of the title molecule due to the substitutions; NH(2) and Cl were deeply investigated by the vibrational analysis. Moreover, (13)C NMR and (1)H NMR chemical shifts were calculated by using the gauge independent atomic orbital (GIAO) method with HF/B3LYP/B3PW91 methods with 6-311++G(d,p). A study on the electronic properties, such as HOMO and LUMO energies, were performed by time-dependent DFT (TD-DFT) approach. Besides frontier molecular orbitals (FMOs), molecular electrostatic potential (MEP) was performed. NLO properties and Mulliken charges of the 2A4CP were also calculated and interpreted. The thermodynamic properties (heat capacity, entropy, and enthalpy) of the title compound at different temperatures were calculated in gas phase.