MilliporeSigma
  • Structural, vibrational, electronic investigations and quantum chemical studies of 2-amino-4-methoxybenzothiazole.
Structural, vibrational, electronic investigations and quantum chemical studies of 2-amino-4-methoxybenzothiazole.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-12)
V Arjunan, Arushma Raj, R Santhanam, M K Marchewka, S Mohan
ABSTRACT
Extensive vibrational investigations of 2-amino-4-methoxybenzothiazole have been carried out with FTIR and FT-Raman spectral techniques. The electronic structure of the molecule has been analysed by UV-Visible and NMR spectroscopies. The DFT studies were carried out with B3LYP and HF methods utilising 6-31G(d,p), 6-311++G(d,p) and cc-pVDZ basis sets to determine the structural, thermodynamical, vibrational, electronic characteristics of the compound and also to understand the electronic and steric influence of the methoxy amino groups on the skeletal frequencies. The mixing of the fundamental modes was determined with the help of total energy distribution (TED). The energies of the frontier molecular orbitals have also been determined. The kinetic and thermodynamic stability and chemical hardness of the molecule have been determined. Complete NBO analysis was also carried out to find out the intramolecular electronic interactions and their stabilisation energy. (1)H and (13)C NMR chemical shifts and the electronic transitions of the molecule are also discussed.