MilliporeSigma
  • Electro-optical tunable waveguide Bragg gratings in lithium niobate induced by femtosecond laser writing.
Electro-optical tunable waveguide Bragg gratings in lithium niobate induced by femtosecond laser writing.
Optics express (2012-11-29)
W Horn, S Kroesen, J Herrmann, J Imbrock, C Denz
ABSTRACT
We report the fabrication of femtosecond laser-induced, first-order waveguide Bragg gratings in lithium niobate in the low repetition rate regime. Type-II waveguides are written into an x-cut lithium niobate wafer and structured periodically to achieve narrowband reflections at wavelengths around 1550 nm. Additionally, electrodes are employed to allow for electro-optic tuning of the spectral response. We demonstrate wavelength control of the central reflection peak by applying a static external electric field. A maximum shift of the reflection peak of Δλ = 625 pm is observed.
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis