MilliporeSigma
  • 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism-related enzymes gene polymorphisms, NNK metabolites levels and urothelial carcinoma.
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism-related enzymes gene polymorphisms, NNK metabolites levels and urothelial carcinoma.
Toxicology letters (2012-11-13)
Chi-Jung Chung, Yeong-Shiau Pu, Horng-Sheng Shiue, Hui-Ling Lee, Pinpin Lin, Hsiu-Yuan Yang, Chien-Tien Su, Yu-Mei Hsueh
ABSTRACT
Gene polymorphisms of the 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism-related enzymes-cytochrome P450 (CYP) monooxygenase 2A13 (CYP2A13) and UDP-glucuronosyltransferases (UGT)-2B7 could contribute to the levels of NNK-related metabolites in urine, thereby increasing the susceptibility to urothelial carcinoma (UC). Therefore, our study aimed to evaluate the roles of two gene polymorphisms (CYP2A13 and UGT2B7) of NNK metabolism-related enzymes in the carcinogenesis of UC in Taiwan. A hospital-based pilot case-control study was conducted. There were 121 UC cases and 121 age- and sex-matched healthy participants recruited from March 2007 to April 2009. Urine samples were analyzed for NNK-related metabolites using the liquid chromatography-tandem mass spectrometry method. Genotyping was conducted using a polymerase chain reaction-restriction fragment length polymorphism technique. ANCOVA and multivariate logistic regression were applied for data analyses. In healthy controls, former smokers had significantly higher total NNAL and higher NNAL-Gluc than never smokers or current smokers. Subjects carrying the UGT2B7 268 His/Tyr or Tyr/Tyr genotype had significantly lower total NNAL than those carrying His/His genotype. However, no association was seen between gene polymorphisms of CYP2A13 and UGT2B7 and UC risk after adjustment for age and sex. Significant dose -response associations between total NNAL, free NNAL, the ratios of free NNAL/total NNAL and NNAL-Gluc/total NNAL and UC risk were observed. In the future, large-scale studies will be required to verify the association between the single nucleotide polymorphisms of NNK metabolism-related enzymes and UC risk.
Product Description
Supelco
4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, analytical standard
Pricing and availability is not currently available.
Supelco
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.