MilliporeSigma
  • Anti-inflammatory effect of sinomenine by inhibition of pro-inflammatory mediators in PMA plus A23187-stimulated HMC-1 Cells.
Anti-inflammatory effect of sinomenine by inhibition of pro-inflammatory mediators in PMA plus A23187-stimulated HMC-1 Cells.
European review for medical and pharmacological sciences (2012-10-11)
Y C Oh, O H Kang, S B Kim, S H Mun, C B Park, Y G Kim, Y I Kim, Y S Lee, S H Han, J H Keum, D W Shin, J Y Ma, D Y Kwon
ABSTRACT
Sinomenine is an alkaloid compound and a prominent anti-inflammatory agent found in the root of the climbing plant Sinomenium acutum. However, its effects on the mechanism of human mast cell line (HMC)-1-mediated inflammation remained unknown. To provide insight into the biological effects of sinomenine, we examined its influence on the pro-inflammatory cytokine production in HMC-1 cells stimulated by phorbol 12-myristate-13-acetate (PMA) plus A23187 by evaluating the stimulated cells in the presence or absence of sinomenine. In the present study, the pro-inflammatory cytokine production was measured using ELISA, Reverse Transcription-polymerase chain reaction (RT-PCR) and nuclear factor (NF)-kappaB, mitogen-activated protein kinases (MAPKs) pathway activation, as determined by Western blot analysis. Also, cyclooxygenase (COX)-2 expression was measured through Western blot and RT-PCR analysis. Sinomenine inhibited the pro-inflammatory cytokine production induced by PMA plus A23187 in a dose-dependent manner. Furthermore, sinomenine inhibited the phosphorylations of extracellular signal-regulated kinase (ERK) and p38 MAPKs as well as the translocation of NF-kappaB p65 through reduced IkappaBalpha degradation. In addition, sinomenine suppressed COX-2 protein and mRNA expression dose-dependently. Taken together, the results of this study indicate that the anti-inflammatory effects of sinomenine may occur via the inhibition of pro-inflammatory cytokine and COX-2 production through the inhibition of MAPKs and NF-kappaB pathway activation by PMA plus A23187 stimulation in HMC-1 cells.