MilliporeSigma
Tm:LiYF4 planar waveguide laser at 1.9 μm.
Optics letters (2012-10-03)
Western Bolanos, Florent Starecki, Abdelmjid Benayad, Gurvan Brasse, Vivien Ménard, Jean-Louis Doualan, Alain Braud, Richard Moncorgé, Patrice Camy
ABSTRACT
Continuous wave laser operation at 1.87 μm of liquid-phase epitaxially (LPE) grown Tm(3+)-doped YLiF(4) (Tm:YLF) layers is demonstrated. The waveguide laser delivers 560 mW by pumping with a Ti:Sapphire laser at 780 nm leading to an efficiency of 76% with respect to the absorbed pump power. This constitutes the first Tm(3+)-doped crystalline fluoride waveguide laser ever demonstrated as well as a record in efficiency and output power for an LPE grown waveguide laser operating in the 2 μm spectral range.