MilliporeSigma
  • Simultaneous enantiomeric determination of MDMA and its phase I and phase II metabolites in urine by liquid chromatography-tandem mass spectrometry with chiral derivatization.
Simultaneous enantiomeric determination of MDMA and its phase I and phase II metabolites in urine by liquid chromatography-tandem mass spectrometry with chiral derivatization.
Analytical and bioanalytical chemistry (2012-09-27)
Keiko Nakanishi, Munehiro Katagi, Kei Zaitsu, Noriaki Shima, Hiroe Kamata, Akihiro Miki, Hajime Kato, Ken-Ichi Harada, Hitoshi Tsuchihashi, Koichi Suzuki
ABSTRACT
A liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) procedure was developed for the simultaneous determination of enantiomers of the prevalent designer drug 3,4-methylenedioxymethamphetamine (MDMA) and its phase I and phase II metabolites in urine with chiral derivatization. The analytes in urine were directly derivatized with chiral Marfey's reagent, N(α)-(5-fluoro-2,4-dinitrophenyl)-D-leucinamide, without extraction. The diastereomers of the N(α)-(2,4-dinitrophenyl)-D-leucinamide derivatives generated were determined by LC-MS/MS. Satisfactory chromatographic separation was achieved for the enantiomers of MDMA and its metabolites 3,4-methylenedioxyamphetamine, 4-hydroxy-3-methoxymethamphetamine (HMMA), HMMA glucuronide, and HMMA sulfate on a semimicro octadecylsilane column using linear gradient elution. With use of multiple reaction monitoring mode, the limits of detection of these analytes ranged from 0.01 to 0.03 μg/mL. Linear calibration curves were obtained for all enantiomers from 0.1 to 20 μg/mL in urine. The method showed sufficient reproducibility and quantitative ability. This is the first report of a simple LC-MS/MS-based analytical procedure with direct chiral derivatization in aqueous media that allows simultaneous enantiomeric determination of drugs and their metabolites, including glucuronide and sulfate derivatives.
Product Description
Supelco
(±)-MDA solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®