MilliporeSigma
  • Phospholipid sources for adrenic acid mobilization in RAW 264.7 macrophages. Comparison with arachidonic acid.
Phospholipid sources for adrenic acid mobilization in RAW 264.7 macrophages. Comparison with arachidonic acid.
Biochimica et biophysica acta (2012-07-25)
Carlos Guijas, Alma M Astudillo, Luis Gil-de-Gómez, Julio M Rubio, María A Balboa, Jesús Balsinde
ABSTRACT
Cells metabolize arachidonic acid (AA) to adrenic acid (AdA) via 2-carbon elongation reactions. Like AA, AdA can be converted into multiple oxygenated metabolites, with important roles in various physiological and pathophysiological processes. However, in contrast to AA, there is virtually no information on how the cells regulate the availability of free AdA for conversion into bioactive products. We have used a comparative lipidomic approach with both gas chromatography and liquid chromatography coupled to mass spectrometry to characterize changes in the levels of AA- and AdA-containing phospholipid species in RAW 264.7 macrophage-like cells. Incubation of the cells with AA results in an extensive conversion to AdA but both fatty acids do not compete with each other for esterification into phospholipids. AdA but not AA, shows preference for incorporation into phospholipids containing stearic acid at the sn-1 position. After stimulation of the cells with zymosan, both AA and AdA are released in large quantities, albeit AA is released to a greater extent. Finally, a variety of phosphatidylcholine and phosphatidylinositol molecular species contribute to AA; however, AdA is liberated exclusively from phosphatidylcholine species. Collectively, these results identify significant differences in the cellular utilization of AA and AdA by the macrophages, suggesting non-redundant biological actions for these two fatty acids.
Sigma-Aldrich
cis-7,10,13,16-Docosatetraenoic acid, ≥98% (GC)