MilliporeSigma
  • Intravenous and oral administrations of DD2 [7-Amino-2-(sulfanylmethyl)heptanoic acid] produce thrombolysis through inhibition of plasma TAFIa in rats with tissue factor-induced microthrombosis.
Intravenous and oral administrations of DD2 [7-Amino-2-(sulfanylmethyl)heptanoic acid] produce thrombolysis through inhibition of plasma TAFIa in rats with tissue factor-induced microthrombosis.
Thrombosis research (2012-07-17)
Tomoyuki Sasaki, Nobuko Yoshimoto, Katsuyoshi Sugimoto, Kimihiko Takada, Norie Murayama, Hiroshi Yamazaki, Keiko Yamamoto, Hidemi Ishii
ABSTRACT
Thrombin-activatable fibrinolysis inhibitor (TAFI) is a plasma zymogen that is activated by thrombin in plasma. In fibrinolytic processes, carboxy-terminal lysine (Lys) residues in partially degraded fibrin are important sites for plasminogen binding and activation, and an active form of TAFI (TAFIa) inhibits fibrinolysis by eliminating these residues proteolytically. We synthesized DD2 [7-Amino-2-(sulfanylmethyl)heptanoic acid], a Lys analogue containing sulfur, as an inhibitor of TAFIa and investigated its pharmacological profile and pathophysiological role in thrombolysis via in vitro and in vivo studies. DD2 specifically inhibited plasma TAFIa activity with an apparent IC(50) (50% inhibitory concentration) value of 3.4×10(-8)M under the present experimental condition and enhanced tissue plasminogen activator-mediated clot lysis in a concentration-dependent manner. In order to study tissue factor (TF)-induced microthrombosis in an animal model, rats were given intravenous injection (2.5mg/kg and higher) or oral administration (10mg/kg and higher) of DD2. This attenuated TF-induced glomerular fibrin deposition and increased the plasma levels of fibrin degradation products and D-dimer in a dose-dependent manner. A DD2 dose approximately 4X higher than the dose used in intravenous injections was required to achieve an equivalent thrombolytic effect to that seen following oral administration. Moreover, the oral absorption efficiency of DD2 into the vasculature was 29.8%. These results indicate that both intravenous and oral administration of DD2 enhanced endogenous fibrinolysis and reduced thrombi in a TF-induced microthrombosis model.
Sigma-Aldrich
7-Aminoheptanoic acid, 98%
Pricing and availability is not currently available.