MilliporeSigma
  • Transcriptome analysis and in vivo activity of fluvastatin versus zoledronic acid in a murine breast cancer metastasis model.
Transcriptome analysis and in vivo activity of fluvastatin versus zoledronic acid in a murine breast cancer metastasis model.
Molecular pharmacology (2012-06-23)
Nadejda Vintonenko, Jean-Philippe Jais, Nadim Kassis, Mohamed Abdelkarim, Gerard-Yves Perret, Marc Lecouvey, Michel Crepin, Melanie Di Benedetto
ABSTRACT
Statins and bisphosphonates are two distinct classes of isoprenoid pathway inhibitors targeting downstream enzyme to HMG-CoA reductase (upstream enzyme) and farnesyl-pyrophosphate synthase, respectively. Here, we studied fluvastatin (Fluva) and zoledronate (Zol), representative molecules of each class, respectively. In vivo metastatic potentials of both molecules were assessed. For the first time, we observed a significant reduction in progression of established metastases with Fluva treatment. Treatment with both Zol at 100 μg/kg and Fluva at 15 mg/kg inhibited 80% of the metastasis bioluminescence signal and increased survival of mice. The Zol and Fluva transcriptomic profiles of treated MDA-MB-231 cells revealed analogous patterns of affected genes, but each of them reached with different kinetics. The observable changes in gene expression started after 24 h for Fluva IC(50 72 h) and only after 48 h for Zol IC(50 72 h). To obtain early changes in gene expression of Zol-treated cells, a 3 times higher dose of Zol IC(50 72 h) had to be applied. Combining Fluva and Zol in vivo showed no synergy, but a benefit of several days in survival of mice. This study demonstrated that Zol or Fluva is of potential clinical use for the treatment of established metastasis.
Product Description
Fluvastatin sodium, European Pharmacopoeia (EP) Reference Standard
Fluvastatin for system suitability, European Pharmacopoeia (EP) Reference Standard