MilliporeSigma
  • Synergistic inhibition of interleukin-6 production in adipose stem cells by tart cherry anthocyanins and atorvastatin.
Synergistic inhibition of interleukin-6 production in adipose stem cells by tart cherry anthocyanins and atorvastatin.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2012-06-19)
Zhou Zhou, Muraleedharan G Nair, Kate J Claycombe
ABSTRACT
Studies have shown positive correlations between inflammatory cytokines such as interleukin-6 (IL-6) and the development of chronic diseases including cardiovascular disease by activating C-reactive protein (CRP). Both atorvastatin calcium (lipitor) as well as flavonoid rich fruit such as tart cherry demonstrate potent anti-inflammatory effects on IL-6 secretion. In this study, we investigated whether tart cherry extract or specific anthocyanins contained in the tart cherry show synergistic anti-inflammatory effects with lipitor. Results showed that LPS-induced adipose stem cell secretion of IL-6 reduced with the addition of tart cherry extract, a mixture of tart cherry anthocyanins, and pure tart cherry cyanidin-3-O-glucoside (C3G) in a dose-dependent manner. Furthermore, lipitor and C3G exhibited synergistic effects in reducing LPS-induced IL-6 secretion from adipose stem cells. In conclusion, these results support potential benefits of using dietary phytochemicals in conjunction with pharmacological therapies to decrease adipose inflammation, drug doses, and ultimately, drug-induced adverse effects.
Sigma-Aldrich
Kuromanin chloride, ≥95% (HPLC)
Pricing and availability is not currently available.