MilliporeSigma
  • Molecular cable-like 1-D iodic spiral chains covered with triple helices stabilized in guest-included chiral porous framework.
Molecular cable-like 1-D iodic spiral chains covered with triple helices stabilized in guest-included chiral porous framework.
Chemical communications (Cambridge, England) (2012-06-14)
Makoto Tadokoro, Yasuko Tanaka, Khoichiro Noguchi, Tomoaki Sugaya, Kyosuke Isoda
ABSTRACT
The supramolecular crystal {[Pr(DMFA)](3)[Ni(II)(Hbim)(3)](2)I}(n) with intricate chiral networks of [Ni(II)(Hbim)(3)](-) molecules is reported. It includes a cationic architecture as a guest, constructed from chiral nanotubes that penetrate I(-) chains with spiral channels wrapped by triple helices. The I(-) chains have AC conductivity in crystals like a molecular cable.