MilliporeSigma
Eight stereoisomers of homonojirimycin from D-mannose.
Organic letters (2012-04-05)
Gabriel M J Lenagh-Snow, Sarah F Jenkinson, Scott J Newberry, Atsushi Kato, Shinpei Nakagawa, Isao Adachi, Mark R Wormald, Akihide Yoshihara, Kenji Morimoto, Kazuya Akimitsu, Ken Izumori, George W J Fleet
ABSTRACT
Although there are 32 6-azidoheptitols, there are only 16 homonojirimycin (HNJ) stereoisomers. Two epimeric azidoalditols derived from d-mannose allow the synthesis in water of eight stereoisomers of HNJ.