Merck
  • Trial of experimental toothpastes regarding quality for cleaning dentures.
Trial of experimental toothpastes regarding quality for cleaning dentures.
The International journal of prosthodontics (2012-03-01)
Ingrid Machado de Andrade, Claudia Helena Silva-Lovato, Raphael Freitas de Souza, Marina Xavier Pisani, Kelly Machado de Andrade, Helena de Freitas Oliveira Paranhos
ABSTRACT
The aim of this study was to evaluate the efficacy of experimental toothpastes for removing denture biofilm by means of a randomized crossover trial. Thirty volunteers brushed their dentures using a brush and four pastes: (1) Corega refreshing mint (control), (2) 0.2% chloramine T, (3) 1.0% chloramine T, and (4) 0.01% fluorosurfactant. Each paste was used for 7 days, and participants were randomized to use them according to one of four sequences. Biofilm was disclosed (neutral red) after each period, photographed, and quantified by means of a software program. All experimental toothpastes were similar to the control in terms of posttreatment biofilm coverage.