MilliporeSigma
  • Effects of one-week versus one-day preoperative treatment with topical 1% prednisolone acetate in dogs undergoing phacoemulsification.
Effects of one-week versus one-day preoperative treatment with topical 1% prednisolone acetate in dogs undergoing phacoemulsification.
Journal of the American Veterinary Medical Association (2012-02-16)
Nancy J McLean, Daniel A Ward, Diane V H Hendrix, Rachel K Vaughn
ABSTRACT
To compare the effects of 2 preoperative anti-inflammatory regimens on intraocular inflammation following phacoemulsification. Randomized controlled trial 21 dogs with immature cataracts. All dogs had cataract surgery via phacoemulsification, and most received prosthetic intraocular lenses. Dogs were randomly divided into 2 groups. Group A dogs were treated topically with prednisolone acetate for 7 days prior to surgery, whereas prednisolone acetate treatment commenced the evening prior to surgery in group B dogs. Postoperative care was identical for both groups. Blood-aqueous barrier breakdown was quantified by use of anterior chamber fluorophotometry, with fluorescein entry into the anterior chamber measured 2 and 9 days after surgery compared with baseline scans obtained prior to surgery. Ophthalmic examinations were performed before surgery and 1 day, 9 days, 3 weeks, 7 weeks, 3 months, and 6 months after surgery. A subjective inflammation score was established at each examination. Intraocular pressures were measured 4 and 8 hours after surgery and at each follow-up examination. There was no difference in the extent of blood-aqueous barrier disruption between the groups at 2 or 9 days after surgery. Subjective inflammation scores were also similar at most time points. Dogs in group A developed postoperative ocular hypertension at a higher frequency (60%) than did those in group B (18%). In dogs that underwent cataract surgery via phacoemulsification, a full week of topical prednisolone acetate treatment prior to surgery did not decrease postoperative inflammation, compared with commencement of topical prednisolone acetate treatment the evening prior to surgery, and was associated with a greater incidence of postoperative ocular hypertension.
Sigma-Aldrich
Prednisolone 21-acetate, ≥97%
Prednisolone acetate, European Pharmacopoeia (EP) Reference Standard