MilliporeSigma
  • Effect of blonanserin on cognitive function in antipsychotic-naïve first-episode schizophrenia.
Effect of blonanserin on cognitive function in antipsychotic-naïve first-episode schizophrenia.
Human psychopharmacology (2012-01-27)
Tomomi Tenjin, Seiya Miyamoto, Nobumi Miyake, Shin Ogino, Rei Kitajima, Kazuaki Ojima, Jun Arai, Haruki Teramoto, Sachiko Tsukahara, Yukie Ito, Masanori Tadokoro, Kiriko Anai, Yasuyuki Funamoto, Yasuhiro Kaneda, Tomiki Sumiyoshi, Noboru Yamaguchi
ABSTRACT
The purpose of this study was to evaluate the effects of blonanserin, a novel antipsychotic, on cognitive function in first-episode schizophrenia. Twenty-four antipsychotic-naïve patients with first-episode schizophrenia participated in the study. Blonanserin was given in an open-label design for 8 weeks. The Brief Assessment of Cognition in Schizophrenia-Japanese language version (BACS-J) was administered as the primary outcome measure at baseline and 8 weeks. Clinical evaluation included the Positive and Negative Syndrome Scale (PANSS), the Schizophrenia Quality of Life Scale-Japanese language version (SQLS-J), and the Clinical Global Impression-Severity of Illness Scale (CGI-S). To exclude the possibility of retest effects on the BACS-J, 10 age-matched patients with chronic schizophrenia treated with blonanserin were tested at baseline and after an 8-week interval. Twenty first-episode patients completed the study. Repeated measures analysis of covariance revealed a significant group-by-time interaction effect on the letter fluency task due to better performance in the first-episode group, but not in the control group. Main effect of time or group-by-time interaction effect on the Tower of London task was not significant; however, the first-episode group, but not the control group, showed substantial improvement with a moderate effect size. All items on the PANSS, SQLS-J, and CGI-S significantly improved after 8 weeks of treatment. These results suggest that blonanserin improves some types of cognitive function associated with prefrontal cortical function.
Sigma-Aldrich
Blonanserin, ≥98% (HPLC)