MilliporeSigma
  • Further characterization of the prototypical nociceptin/orphanin FQ peptide receptor agonist Ro 64-6198 in rodent models of conflict anxiety and despair.
Further characterization of the prototypical nociceptin/orphanin FQ peptide receptor agonist Ro 64-6198 in rodent models of conflict anxiety and despair.
Psychopharmacology (2012-01-18)
Celia Goeldner, Will Spooren, Jürgen Wichmann, Eric P Prinssen
ABSTRACT
Ro 64-6198, the prototypical non-peptide nociceptin/orphanin FQ peptide (NOP) receptor agonist, has potent anxiolytic-like effects in several preclinical models and species. However the effects of Ro 64-6198 on distinctive anxiety-provoking conditions related to unconditioned conflict behavior as well as its role in despair-like behavior remain to be addressed. Here we examined the effects of Ro 64-6198 on unconditioned conflict anxiety using stimuli with different salience and on regulation of autonomic reactivity and compared these to the effects of benzodiazepine receptor agonists. We also addressed the potential effects of Ro 64-6198 on despair-like behavior. Ro 64-6198 (0.1 to 10 mg/kg i.p.) and either diazepam or chlordiazepoxide were tested in the Vogel conflict punished drinking test (VCT) in Sprague Dawley rats, in the social approach-avoidance (SAA) test in Lewis rats, in the novelty-induced hypophagia (NIH) in C57BL/6J mice, and in stress-induced hyperthermia in NMRI mice, as well as in the forced swim test (FST) in Sprague Dawley rats and the tail suspension test (TST) in C57BL/6J mice. Ro 64-6198 (0.3 to 3 mg/kg) dose-dependently produced anxiolytic-like effects in the VCT, SAA, NIH, and SIH, similar to benzodiazepine receptor agonists. Ro 64-6198 did not alter immobility time in the FST and TST. Ro 64-6198 produced marked anxiolytic-like effects in response to a variety of mild to strong anxiogenic stimuli, whereas it did not facilitate depression-related behaviors. This data extend previous literature suggesting that NOP receptors are a viable target for the treatment of anxiety disorders.
Sigma-Aldrich
Ro 64-6198, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Chlordiazepoxide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.