MilliporeSigma
  • [Dentin bonding of cements. The bonding of cements with dentin in combination with various indirect restorative materials].
[Dentin bonding of cements. The bonding of cements with dentin in combination with various indirect restorative materials].
Schweizer Monatsschrift fur Zahnmedizin = Revue mensuelle suisse d'odonto-stomatologie = Rivista mensile svizzera di odontologia e stomatologia (2011-12-29)
Anne Peutzfeldt, Alireza Sahafi, Simon Flury
ABSTRACT
The number of both luting agents and restorative materials available on the market has rapidly increased. This study compared various types of luting agents when used to bond different indirect, laboratory restorative materials to dentin. Cylinders were produced of six restorative materials (gold alloy, titanium, feldspathic porcelain, leucite-glass ceramic, zirconia, and an indirect resin composite). Following relevant pretreatment, the end surface of the cylinders were luted to ground, human dentin with eight different luting agents (DeTrey Zinc [zinc phosphate cement], Fuji I [conventional glass ionomer cement], Fuji Plus [resin-modified glass ionomer cement], Variolink II [conventional etch-and-rinse resin cement], Panavia F2.0 and Multilink [self-etch resin cements], RelyX Unicem Aplicap and Maxcem [self-adhesive resin cements]). After water storage at 37 °C for one week, the shear bond strength of the specimens was measured and the fracture mode was examined stereo-microscopically. Restorative material and luting agent both had a significant effect on bond strength and there was a significant interaction between the two variables. The zinc phosphate cement and the glass ionomer cements resulted in the lowest bond strengths, whereas the highest bond strengths were found with the two self-etch and one of the self-adhesive resin cements.
Sigma-Aldrich
Zinc phosphate, 99.998% trace metals basis