MilliporeSigma
  • Lycorine inhibits lipopolysaccharide-induced iNOS and COX-2 up-regulation in RAW264.7 cells through suppressing P38 and STATs activation and increases the survival rate of mice after LPS challenge.
Lycorine inhibits lipopolysaccharide-induced iNOS and COX-2 up-regulation in RAW264.7 cells through suppressing P38 and STATs activation and increases the survival rate of mice after LPS challenge.
International immunopharmacology (2011-12-14)
Jingjing Kang, Yushun Zhang, Xiang Cao, Jie Fan, Guilan Li, Qi Wang, Ying Diao, Zhihui Zhao, Lan Luo, Zhimin Yin
ABSTRACT
As a natural alkaloid extracted from Amaryllidaceae, lycorine shows various biological effects on tumor cells. Here we show that lycorine dose-dependently inhibited the LPS-induced up-regulation of nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) protein level in RAW264.7 cells. Besides, it also inhibited NO, PGE(2), TNF-α and IL-6 release from LPS-treated RAW264.7 cells. RT-PCR experiments showed that lycorine suppressed LPS-induced iNOS but not COX-2 gene expression. Moreover, lycorine decreased LPS-induced mortality in mice. Mechanistically, LPS-induced activation of P38 and STATs pathways was suppressed significantly by lycorine. In addition, lycorine did not interfere with the phosphorylation of ERK1/2, JNK1/2 and NF-κB pathways. In conclusion, lycorine inhibits LPS-induced production of pro-inflammatory mediators and increases the survival rate of mice after LPS challenge, suggesting that lycorine could play an anti-inflammatory role in response to LPS.
Sigma-Aldrich
Lycorine hydrochloride, ≥98% (TLC), powder