MilliporeSigma
  • A newly high alkaline lipase: an ideal choice for application in detergent formulations.
A newly high alkaline lipase: an ideal choice for application in detergent formulations.
Lipids in health and disease (2011-11-30)
Slim Cherif, Sami Mnif, Fatma Hadrich, Slim Abdelkafi, Sami Sayadi
ABSTRACT
Bacterial lipases received much attention for their substrate specificity and their ability to function in extreme environments (pH, temperature...). Many staphylococci produced lipases which were released into the culture medium. Reports of thermostable lipases from Staphylococcus sp. and active in alkaline conditions are not previously described. A newly soil-isolated Staphylococcus sp. strain ESW secretes an induced lipase in the culture medium. The effects of temperature, pH and various components in a detergent on the activity and stability of Staphylococcus sp. lipase (SL1) were studied in a preliminary evaluation for use in detergent formulation solutions. The enzyme was highly active over a wide range of pH from 9.0 to 13.0, with an optimum at pH 12.0. The relative activity at pH 13.0 was about 60% of that obtained at pH 12.0. It exhibited maximal activity at 60°C. This novel lipase, showed extreme stability towards non-ionic and anionic surfactants after pre-incubation for 1 h at 40°C, and relative stability towards oxidizing agents. Additionally, the crude enzyme showed excellent stability and compatibility with various commercial solid and liquid detergents. These properties added to the high activity in high alkaline pH make this novel lipase an ideal choice for application in detergent formulations.