MilliporeSigma
  • Facile synthesis of AgBr nanoplates with exposed {111} facets and enhanced photocatalytic properties.
Facile synthesis of AgBr nanoplates with exposed {111} facets and enhanced photocatalytic properties.
Chemical communications (Cambridge, England) (2011-11-23)
Hua Wang, Jian Gao, Tianqi Guo, Rongming Wang, Lin Guo, Yang Liu, Jinghong Li
ABSTRACT
AgBr nanoplates with exposed {111} facets have been synthesized in high yield by a facile precipitation reaction, and the as-prepared nanoplates exhibited greatly enhanced photocatalytic properties for the degradation of organic pollutants, which may be primarily ascribed to the relatively higher surface energy of {111} facets.
Sigma-Aldrich
Silver bromide, 99%