MilliporeSigma
Ginsenoside Re: pharmacological effects on cardiovascular system.
Cardiovascular therapeutics (2011-09-03)
Lu Peng, Shi Sun, Lai-Hua Xie, Sheila M Wicks, Jing-Tian Xie
ABSTRACT
Ginsenosides are the bioactive constituents of ginseng, a key herb in traditional Chinese medicine. As a single component of ginseng, ginsenoside Re (G-Re) belongs to the panaxatriol group. Many reports demonstrated that G-Re possesses the multifaceted beneficial pharmacological effects on cardiovascular system. G-Re has negative effect on cardiac contractility and autorhythmicity. It causes alternations in cardiac electrophysiological properties, which may account for its antiarrhythmic effect. In addition, G-Re also exerts antiischemic effect and induces angiogenic regeneration. In this review, we first outline the chemistry and the pharmacological effects of G-Re on the cardiovascular system.
Product Description
Supelco
Ginsenoside Re, analytical standard
Ginsenoside Re, primary reference standard