MilliporeSigma
  • New transposon Tn6133 in methicillin-resistant Staphylococcus aureus ST398 contains vga(E), a novel streptogramin A, pleuromutilin, and lincosamide resistance gene.
New transposon Tn6133 in methicillin-resistant Staphylococcus aureus ST398 contains vga(E), a novel streptogramin A, pleuromutilin, and lincosamide resistance gene.
Antimicrobial agents and chemotherapy (2011-07-20)
Sybille Schwendener, Vincent Perreten
ABSTRACT
A novel streptogramin A, pleuromutilin, and lincosamide resistance determinant, Vga(E), was identified in porcine methicillin-resistant Staphylococcus aureus (MRSA) ST398. The vga(E) gene encoded a 524-amino-acid protein belonging to the ABC transporter family. It was found on a multidrug resistance-conferring transposon, Tn6133, which was comprised of Tn554 with a stably integrated 4,787-bp DNA sequence harboring vga(E). Detection of Tn6133 in several porcine MRSA ST398 isolates and its ability to circularize suggest a potential for dissemination.
Sigma-Aldrich
Pleuromutilin, ≥95%