MilliporeSigma
A prototype antifungal contact lens.
Investigative ophthalmology & visual science (2011-04-30)
Joseph B Ciolino, Sarah P Hudson, Ashley N Mobbs, Todd R Hoare, Naomi G Iwata, Gerald R Fink, Daniel S Kohane
ABSTRACT
To design a contact lens to treat and prevent fungal ocular infections. Curved contact lenses were created by encapsulating econazole-impregnated poly(lactic-co-glycolic) acid (PLGA) films in poly(hydroxyethyl methacrylate) (pHEMA) by ultraviolet photopolymerization. Release studies were conducted in phosphate-buffered saline at 37°C with continuous shaking. The contact lenses and their release media were tested in an antifungal assay against Candida albicans. Cross sections of the pre- and postrelease contact lenses were characterized by scanning electron microscopy and by Raman spectroscopy. Econazole-eluting contact lenses provided extended antifungal activity against Candida albicans fungi. Fungicidal activity varied in duration and effectiveness depending on the mass of the econazole-PLGA film encapsulated in the contact lens. An econazole-eluting contact lens could be used as a treatment for fungal ocular infections.
Product Description
Econazole nitrate, European Pharmacopoeia (EP) Reference Standard
Econazole, European Pharmacopoeia (EP) Reference Standard