MilliporeSigma
  • Synthesis of the marine bromotyrosine psammaplin F and crystal structure of a psammaplin A analogue.
Synthesis of the marine bromotyrosine psammaplin F and crystal structure of a psammaplin A analogue.
Molecules (Basel, Switzerland) (2010-12-04)
Qianjiao Yang, Dan Liu, Deyang Sun, Sen Yang, Guodong Hu, Zuti Wu, Linxiang Zhao
ABSTRACT
Psammaplin F, an unsymmetrical disulfide bromotyrosine, was isolated from the sponge Pseudoceratina purpurea in 2003. We reported here the first total synthesis of psammaplin F in 12% overall yield by employing Cleland's reagent reduction as key step. The longest linear synthetic sequence starting from 3-bromo-4-hydroxybenzaldehyde and hydantoin was seven steps. In addition, a detailed X-ray crystal structure analysis of psammaplin A analogue 8b is given for the first time.
Sigma-Aldrich
Cystamine dihydrochloride, 96%