MilliporeSigma
  • Excited-state prototropic equilibrium dynamics of 6-hydroxyquinoline encapsulated in microporous catalytic faujasite zeolites.
Excited-state prototropic equilibrium dynamics of 6-hydroxyquinoline encapsulated in microporous catalytic faujasite zeolites.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-09-21)
Sun-Young Park, Hyunung Yu, Jiho Park, Du-Jeon Jang
ABSTRACT
The excited-state proton transfer and geminate recombination of 6-hydroxyquinoline (6HQ) encaged in catalytic Na(+)-exchanged faujasite zeolites X (NaX) and Y (NaY) have been explored by measuring steady-state and picosecond time-resolved spectra. The pathways and rate constants of proton transfer of excited 6HQ are determined by the microscopic environment of zeolitic hosts surrounding the guest molecules. The excited-state proton transfer of a 6HQ molecule encapsulated in a zeolitic nanocavity is initiated by deprotonation of the enolic group to form an anionic intermediate and completed by subsequent protonation of the imino group to form a zwitterionic tautomer. Geminate recombination occurs to compete with proton transfer at each tautomerization step of excited-state 6HQ because of the confined environment of dehydrated zeolitic supercages. Consequently, excited-state equilibria among three prototropic species of 6HQ are established in microporous catalytic faujasite zeolites. Kinetic differences in NaX and NaY are attributed to dissimilarities in acidity/basicity.
Sigma-Aldrich
6-Hydroxyquinoline, 95%