MilliporeSigma
  • A single-blind, comparative study of zotepine versus haloperidol in combination with a mood stabilizer for patients with moderate-to-severe mania.
A single-blind, comparative study of zotepine versus haloperidol in combination with a mood stabilizer for patients with moderate-to-severe mania.
Psychiatry and clinical neurosciences (2010-05-08)
Hung-Yu Chan, Shaw-Hua Jou, Yeong-Yuh Juang, Ching-Jui Chang, Jiahn-Jyh Chen, Chiung-Hsu Chen, Nan-Ying Chiu
ABSTRACT
Atypical antipsychotics are increasingly used in the management of acute mania. This study was conducted to investigate the efficacy and tolerability of zotepine compared to haloperidol in combination with a mood stabilizer (lithium or valproate) for treatment of acute mania. This was a multi-center, randomized, rater-blinded, parallel-group, flexible-dose study. Forty-five hospitalized patients with moderate-to-severe manic, bipolar disorder (DSM-IV) were randomly assigned to a zotepine or a haloperidol 4-week treatment group. There was no significant between-group difference in the Young Mania Rating Scale total scores between the zotepine and haloperidol groups (-23.7 + or - 12.1 vs -22.3 + or - 11.0, respectively). The adverse events in both groups were mild to moderate. The haloperidol group reported a higher incidence of treatment-related adverse events, especially parkinsonism and akathisia, compared to the zotepine group. Serum uric acid decreased more in the zotepine group than in the haloperidol group. In combination with a mood stabilizer, zotepine appears to be as effective as haloperidol in treating moderate-to-severe mania in the acute phase, but has the advantages of lowering hyperuricemia and fewer extrapyramidal side-effects. Double-blinded studies with larger sample sizes are warranted to confirm these findings.
Sigma-Aldrich
Zotepine