MilliporeSigma
  • Structural basis for the efficient phosphorylation of AZT-MP (3'-azido-3'-deoxythymidine monophosphate) and dGMP by Plasmodium falciparum type I thymidylate kinase.
Structural basis for the efficient phosphorylation of AZT-MP (3'-azido-3'-deoxythymidine monophosphate) and dGMP by Plasmodium falciparum type I thymidylate kinase.
The Biochemical journal (2010-04-01)
Jean L Whittingham, Juana Carrero-Lerida, James A Brannigan, Luis M Ruiz-Perez, Ana P G Silva, Mark J Fogg, Anthony J Wilkinson, Ian H Gilbert, Keith S Wilson, Dolores González-Pacanowska
ABSTRACT
Plasmodium falciparum is the causative agent of malaria, a disease where new drug targets are required due to increasing resistance to current anti-malarials. TMPK (thymidylate kinase) is a good candidate as it is essential for the synthesis of dTTP, a critical precursor of DNA and has been much studied due to its role in prodrug activation and as a drug target. Type I TMPKs, such as the human enzyme, phosphorylate the substrate AZT (3'-azido-3'-deoxythymidine)-MP (monophosphate) inefficiently compared with type II TMPKs (e.g. Escherichia coli TMPK). In the present paper we report that eukaryotic PfTMPK (P. falciparum TMPK) presents sequence features of a type I enzyme yet the kinetic parameters for AZT-MP phosphorylation are similar to those of the highly efficient E. coli enzyme. Structural information shows that this is explained by a different juxtaposition of the P-loop and the azide of AZT-MP. Subsequent formation of the transition state requires no further movement of the PfTMPK P-loop, with no steric conflicts for the azide moiety, allowing efficient phosphate transfer. Likewise, we present results that confirm the ability of the enzyme to uniquely accept dGMP as a substrate and shed light on the basis for its wider substrate specificity. Information resulting from two ternary complexes (dTMP-ADP and AZT-MP-ADP) and a binary complex with the transition state analogue AP5dT [P1-(5'-adenosyl)-P5-(5'-thymidyl) pentaphosphate] all reveal significant differences with the human enzyme, notably in the lid region and in the P-loop which may be exploited in the rational design of Plasmodium-specific TMPK inhibitors with therapeutic potential.
Sigma-Aldrich
2′-Deoxyguanosine 5′-monophosphate disodium salt hydrate, ≥98%