MilliporeSigma
  • Reaction of N-hydroxyguanidine with the ferrous-oxy state of a heme peroxidase cavity mutant: a model for the reactions of nitric oxide synthase.
Reaction of N-hydroxyguanidine with the ferrous-oxy state of a heme peroxidase cavity mutant: a model for the reactions of nitric oxide synthase.
Archives of biochemistry and biophysics (2010-03-30)
Alycen Pond Nigro, David B Goodin
ABSTRACT
Yeast cytochrome c peroxidase was used to construct a model for the reactions catalyzed by the second cycle of nitric oxide synthase. The R48A/W191F mutant introduced a binding site for N-hydroxyguanidine near the distal heme face and removed the redox active Trp-191 radical site. Both the R48A and R48A/W191F mutants catalyzed the H2O2 dependent conversion of N-hydroxyguanidine to N-nitrosoguanidine. It is proposed that these reactions proceed by direct one-electron oxidation of NHG by the Fe(+4)O center of either Compound I (Fe(+4)=O, porph+(.)) or Compound ES (Fe(+4)=O, Trp+(.)). R48A/W191F formed a Fe(+2)O2 complex upon photolysis of Fe(+2)CO in the presence of O2, and N-hydroxyguanidine was observed to react with this species to produce products, distinct from N-nitrosoguanidine, that gave a positive Griess reaction for nitrate+nitrite, a positive Berthelot reaction for urea, and no evidence for formation of NO(.). It is proposed that HNO and urea are produced in analogy with reactions of nitric oxide synthase in the pterin-free state.
Sigma-Aldrich
Hydroxyguanidine sulfate salt