MilliporeSigma
  • Influence of the metal complex-to-peptide linker on the synthesis and properties of bioactive CpMn(CO)3 peptide conjugates.
Influence of the metal complex-to-peptide linker on the synthesis and properties of bioactive CpMn(CO)3 peptide conjugates.
Dalton transactions (Cambridge, England : 2003) (2010-02-25)
Katrin Splith, Ines Neundorf, Wanning Hu, Harmel W Peindy N'Dongo, Vera Vasylyeva, Klaus Merz, Ulrich Schatzschneider
ABSTRACT
By combining organometallic groups and peptides, a large number of conjugates with interesting new biological properties can be prepared. Especially, attachment to cell-penetrating peptides (CPP) that act as efficient cell delivery vehicles has come to the fore. However, the presence of the metal moiety in such systems can interfere with standard conjugate synthesis procedures which therefore need to be optimized for every new compound. In this work, we report on the preparation of six new cymantrene-sC18 peptide bioconjugates that were prepared by solid phase peptide synthesis (SPPS) techniques. The cymantrene complexes were chosen for their different linker to the peptide, to study the influence of the linker group on cellular uptake and cell viability of the conjugates. Interestingly, the attachment of the metal complex leads to a non-standard cleavage of the Rink amide linker used in the SPPS protocol under trifluoroacetic acid (TFA) treatment, resulting in peptide amides that are N-alkylated at the C-terminus. Furthermore, we found that depending on the type of cymantrene moiety attached, the formation of reactive carbocations which result from decomposition of the resin linker is facilitated and can alkylate the metal complex moiety. Both effects were analyzed by MS/MS studies and cleavage mixtures for efficient elimination of this byproduct formation were identified. Moreover, initial biological testing of the cytotoxicity of one of the bioconjugates gave promising results. Concentration-dependent cell viability studies of Cym1-sC18 on human MCF-7 breast adenocarcinoma cells gave an IC(50) value of 59.8 (+/- 6.7) microM and demonstrate their potential in anticancer chemotherapy.
Sigma-Aldrich
Cyclopentadienylmanganese(I) tricarbonyl