MilliporeSigma
  • The pan-ErbB receptor tyrosine kinase inhibitor canertinib induces ErbB-independent apoptosis in human leukemia (HL-60 and U-937) cells.
The pan-ErbB receptor tyrosine kinase inhibitor canertinib induces ErbB-independent apoptosis in human leukemia (HL-60 and U-937) cells.
Biochemical and biophysical research communications (2010-01-26)
Cecilia Trinks, Emelie A Djerf, Anna-Lotta Hallbeck, Jan-Ingvar Jönsson, Thomas M Walz
ABSTRACT
Epidermal growth factor (EGF) receptor tyrosine kinase inhibitors have recently been shown to display anti-neoplastic effects in human malignant myeloid cells. Our study was initiated in order to determine the effect of the pan-ErbB receptor tyrosine kinase inhibitor, canertinib (CI-1033), on growth and survival of human leukemia (HL-60 and U-937) cells. We show that treatment of HL-60 and U-937 cells with canertinib significantly inhibits growth of both cell lines in a dose-dependent manner; half maximal effective dose (IC(50)) in HL-60 and U-937 cells was approximately 2.5 microM and 1.0 microM, respectively. Treatment with 2 microM canertinib promoted a G(1) cell cycle arrest, whereas doses of 5 microM or more induced apoptosis as determined by the Annexin V method and cleavage of poly-(ADP-ribose) polymerase (PARP). HL-60 and U-937 cells lacked EGF-receptor transcript but expressed ErbB2-4 mRNA as determined by RT-PCR. However, none of the corresponding ErbB-receptor proteins could be detected by Western blot analysis. We conclude that canertinib induces apoptosis in HL-60 and U-937 cells devoid of functional ErbB1-4 receptors. Our results suggest that canertinib could be of potential clinical interest in the treatment of acute myeloid leukemia.
Sigma-Aldrich
CI-1033, ≥98% (HPLC)