MilliporeSigma
  • Changes in N-acetylglutamate are involved in regulating urea synthesis in rats given a low gluten diet supplemented with L-lysine, L-methinone and L-threonine.
Changes in N-acetylglutamate are involved in regulating urea synthesis in rats given a low gluten diet supplemented with L-lysine, L-methinone and L-threonine.
Journal of nutritional science and vitaminology (2009-11-21)
Kazuyo Tujioka, Tamami Tuchiya, Xianglan Shi, Miho Ohsumi, Kazutoshi Hayase, Hidehiko Yokogoshi
ABSTRACT
We have shown that urinary urea excretion decreased in rats fed a low gluten diet supplemented with dietary limiting amino acids. The purpose of present study was to determine whether the addition of dietary limiting amino acids to a low gluten diet affected the synthesis and degradation of N-acetylglutamate and regulated urea synthesis. Experiments were done on two groups of rats, given diets containing 10% gluten or 10% gluten+0.5% L-lysine, 0.2% L-threonine and 0.2% L-methionine for 10 d. The urinary excretion of urea, and the liver concentration of N-acetylglutamate, and the liver activity of N-acetylglutamate synthetase decreased with the addition of dietary L-lysine, L-threonine and L-methionine. N-Acetylglutamate concentration in the liver was closely correlated with the N-acetylglutamate synthetase activity in the liver and excretion of urea. The greater degradation of N-acetylglutamate was observed in the group fed the 10% gluten+L-lysine, L-threonine and L-methionine. The hepatic concentration of glutamate and plasma concentration of arginine were not related to the N-acetylglutamate concentration in the liver. These results suggest that the addition of limiting amino acids to the low gluten diet controls the synthesis and degradation of N-acetylglutamate in the liver and lowers urea synthesis.