MilliporeSigma
  • Infusions of albumin increase free fraction of naproxen in healthy volunteers: a randomized crossover study.
Infusions of albumin increase free fraction of naproxen in healthy volunteers: a randomized crossover study.
Acta anaesthesiologica Scandinavica (2009-11-03)
P A Reine, U E Kongsgaard, A Andersen, A-K Thøgersen, H Olsen
ABSTRACT
In vitro and in vivo studies have indicated that stabilizers present in pharmaceutical-grade albumin influence the albumin-binding capacity for highly protein-bound drugs. However, the half-life of the stabilizers and the quantitative effect have been difficult to determine. A randomized crossover study including six healthy volunteers was performed. The study subjects received 750 mg of oral naproxen 2 h before the study. They were randomized to receive either 100 ml of 20% albumin or 100 ml of Ringer's acetate solution intravenously. Frequent blood samples were obtained. The experiment was repeated 4 weeks later with the alternate solution. The serum samples were analysed to determine the concentrations of albumin, N-acetyl-DL-tryptophan, caprylate, and naproxen. The free fraction of naproxen increased significantly after the infusion of albumin (P<0.05). The increase was concurrent with the appearance of N-acetyl-DL-tryptophan and caprylate in serum. The free fraction of naproxen declined rapidly after the albumin infusion was completed. N-acetyl-DL-tryptophan had a half-life of approximately 30 min. The half-life of caprylate was <15 min. A transfusion of albumin results in an increase in the free fraction of naproxen. The transient increase in free-fraction naproxen decreased together with the detectable levels of the stabilizers N-acetyl-DL-tryptophan and caprylate. N-acetyl-DL-tryptophan and caprylate have a short half-life in serum.
Product Description
Supelco
N-Acetyl-L-tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-tryptophan